GE Druck PTX5072-TC-A2-CA-H6-PE High-Accuracy Pressure Transmitter, 5000 psi, Sealed

Representative image only.
GE Druck Mfr # PTX5072-TC-A2-CA-H6-PE – Item # EW-98071-10
All-stainless-steel construction for heavy industrial use
  • ±0.1% full-scale accuracy
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Accuracy±0.1% full-scale (combined effects of nonlinearity, hysteresis, and repeatability)
  • Min Pressure (PSI)0
  • Max Pressure (PSI)5000
  • Wetted Materials316L SS, Hastelloy C276
  • Process Connection1/4\" NPT(F)
  • Battery Type7 to 28 VDC (Note: supply voltage is 7 to 32 VDC in nonhazardous area operation)
  • Electrical ConnectionsDIN 43650 Form A demountable (mating connector supplied)
  • Output4 to 20 mA
  • Media CompatibilityGases and liquids compatible with wetted materials
  • DescriptionHigh-Accuracy Pressure Transmitter, 5000 psi, Sealed
  • Warranty1 year
1 Year WarrantyConformite EuropeenneFM ApprovedIntrinsically Safe

Key Features

  • ±0.1% full-scale accuracy
  • 1 year warranty

More About this Item

These high-accuracy transmitters combine proven silicon technology and analog circuitry with a compact design. The Druck silicon technology is at the heart of the transmitter, resulting in a range of high quality piezo-resistive pressure sensors which are both robust and stable. High levels of performance have been achieved without the use of digital electronics. These transmitters are particularly well suited to sensitive applications, such as those requiring low power, low noise, pulse power, and fast response.

High-grade stainless steel body and analog electronics provide fast turn on, pulse power, 5 kHz frequency response, and low noise. Additionally, transmitters feature high over-pressure capability, stability typically at ±0.05%, and FM (US and Canada) intrinsically safe certifications.

Higher pressure range transmitters come standard with sealed diaphragm to provide extra pressure equipment safety. Other process and electrical connections are available; contact an Application Specialist for details.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat