GE Druck PV 621G Pneumatic Pressure Station 300 psi
Representative image only.
GE Druck Mfr # PV621G – Item # EW-23000-87
Acts as a pressure source for the DPI620G or as a stand-alone pressure generator
  • 95% vacuum to 300 psi pneumatic
  • 1 year warranty
$2,203.00USD / Each
sign in for your price.
$227.00USD / Each
for an additional
$227.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure300 psi (20 bar)
  • DescriptionPneumatic Pressure Station 300 psi
  • Warranty1 Year
1 Year WarrantyIP 65
Key Features
  • 95% vacuum to 300 psi pneumatic
  • 1 year warranty
More About this Item
The PV 621 generates pressure for use with the DPI620G or as a stand alone pressure generator. When combined with the DPI620G and one of many pressure modules, it forms a fully-integrated pressure calibrator of unparalleled performance.
As a stand-alone pressure station it replaces conventional hand pumps providing greater efficiency and significantly reducing the risk of repetitive strain injuries. The PV 621 generates pressures up to 300 psi (20 bar) using pneumatic noncorrosive gases.