GE Druck PV 623G Pressure Station 15000 psi
Representative image only.
GE Druck Mfr # PV623G – Item # EW-23000-89
Acts as a pressure source for the DPI620G or as a stand-alone pressure generator
  • 0 to 15,000 psi hydraulic
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure15,000 psi (1000 bar)
  • DescriptionPressure Station 15000 psi
  • Warranty1 Year
1 Year WarrantyIP 65
Key Features
  • 0 to 15,000 psi hydraulic
  • 1 year warranty
More About this Item
The PV 623 generates pressure for use with the DPI620G or as a stand alone pressure generator. When combined with the DPI620G and one of many optional pressure modules, it forms a fully-integrated pressure calibrator of unparalleled performance.
As a stand-alone pressure station it replaces conventional hand pumps providing greater efficiency and significantly reducing the risk of repetitive strain injuries. The PV 623 generates pressures up to 15,000 psi (1000 bar) using demineralized water or mineral oil.