GE Druck PV411 104 Pump & 5,000 PSIG Gauge w/Hose, Adapters, & Case
Representative image only.
GE Druck Mfr # PV411-104-HP-2-20G – Item # EW-68437-32
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPump & 5,000 PSIG Gauge w/Hose, Adapters, & Case
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The DPI 104 is a micro-processor controlled digital pressure gauge that combines precision and functionality in a compact, robust and simple to use package. The DPI 104 matches advanced silicon sensor technology with several convenient design features resulting in an accurate, versatile yet affordable digital test gauge. Supplied as a stand-alone process indicator or in a kit with the widely proven Druck hand pumps, the DPI 104 provides a reliable and economic solution for a wide range of pressure sensing applications.
FEATURES
  • 0.05% full scale (FS) accuracy
  • Pressure ranges to 20,000 psi (1400 bar)
  • Eleven selectable pressure units
  • Large, easy to read display with five digit resolution
  • % pressure indication and additional bar graph for quick visual reference
  • Temperature compensated accuracy from 14°F to 122°F (-10°C to 50°C)
  • 0 to 5V analog output
  • Pressure switch test
  • Minimum/Maximum, tare and alarm functions
  • IDOS compatible and RS232 serial interface
  • Networking capability (1 to 99 units)
  • Stainless steel or Inconel pressure cavity for aggressive media
APPLICATIONS
  • Process monitoring and control
  • Test and calibration