GE Druck PV411A Hand Pump 0/10000PSI Hydraulic 0/10000PSI Generation
Representative image only.
GE Druck Mfr # PV411A – Item # EW-68437-48
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHand Pump 0/10000PSI Hydraulic 0/10000PSI Generation
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The PV411A provides functionality never before seen in a hand held pressure source. It can replace four conventional hand pumps.
FEATURES
  • Vacuum priming function to remove air during hydraulic operation
  • High pressure generation accomplished with volume adjuster rather than physician effort
  • Tamper-proof adjustable overpressure protection valve
  • 240° swivel head for optimum viewing of reference indicators
  • Pressure/vacuum selector provides fine control in both pneumatic and hydraulic modes
  • High sensitivity volume adjuster for hydraulic, pneumatic, vacuum and low pressure operation
  • Built-in stroke adjuster controls pressure / vacuum generated per stroke
  • PTFE anodized internal moving parts for reduced friction and wear
  • Removable hydraulic reservoir can be disconnected from pump without draining
The GE411AHA Hand Pump Kit includes heavy-duty transport case with closed foam insert, 10,000 PSI high pressure hose assembly 3 ft length, 2-1/4" male NPT adapters, 1-1/4" female NPT adapter, 1-1/8" female NPT adapter and 1-1/4" male to 1/8" female adapter.