GE Panametrics C-RS-402-00-0 Transducer Pair, for 2 to 24" Pipe, 1.0MHz

Representative image only.
GE Panametrics Mfr # C-RS-402-00-0 – Item # EW-68453-02

$1,710.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Compatible Withliquids
  • Length (in)9.4
  • Width (in)5.5
  • Width (cm)14
  • Length (cm)23.9
  • Width (mm)140
  • Length (mm)239
  • DescriptionTransducer Pair, for 2 to 24" Pipe, 1.0MHz

More About this Item

Keep your GE Panametrics flowmeter running accurately with these parts and accessories.

Available for sale in the United States and Canada only.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat