This item is now discontinued. Please browse similar items
GE Panametrics PT9-BNCAdapt TNC to BNC Adapter for Existing Transducers
Representative image only.
GE Panametrics Mfr # PT9-BNCAdapt – Item # EW-24500-84
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithPT878 transducers and PT900 ultrasonic flowmeter.
  • ConnectionsBNC male to TNC female
  • DescriptionTNC to BNC Adapter for Existing Transducers
More About this Item
Use existing transducers from the previous version (PT878) with the new PT900 ultrasonic flowmeter.