GE SI Pressure Instruments PV210 Low-Pressure and Vacuum Hand Pump, 0-45 PSI
Representative image only.
GE SI Pressure Instruments Mfr # PV210 – Item # EW-68974-30
Portable hand pump in lighweight yet rugged and durable
  • Vacuum to -27 inHg
  • Precise fine control to 0.001" H2O
  • Pneumatic pressures to 1250" H2O/45 psi
$669.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure45 psi
  • Min Pressure27 inHg
  • Height (in)6 3/4
  • Diameter (in)1 3/4
  • DescriptionLow-Pressure and Vacuum Hand Pump, 0-45 PSI
Includes
two flexible 3-ft, 1/8"-dia hoses with 1/4" NPT(F) adapters
Key Features
  • Vacuum to -27 inHg
  • Precise fine control to 0.001" H2O
  • Pneumatic pressures to 1250" H2O/45 psi
More About this Item
Thermally-insulated to eliminate temperature effect, this test pump is designed for the requirements of low pressure calibration. It is a dual source for pneumatic pressure and vacuum, and features finger-tight quick-fit connectors, a built-in pressure relief valve, and a fine control needle valve for precise pressure release.