GE Druck PV212 Hydraulic Hand Pump; 10,000 psi
Representative image only.
GE Druck Mfr # PV212-10K – Item # EW-68974-34
Pressure relief valve offers outstanding control for deacreasing pressures
  • Generate pressure to 10,000 psi
  • Vernier adjustment for fine control
  • Quick priming and pressure generation
  • 18 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure10000 psi
  • Min Pressure0 psi
  • DescriptionHydraulic Hand Pump; 10,000 psi
  • Warranty18 Months
18 Month Warranty
Key Features
  • Generate pressure to 10,000 psi
  • Vernier adjustment for fine control
  • Quick priming and pressure generation
  • 18 month warranty
More About this Item
Hydraulic hand pump includes a selector valve that increases the priming speed and greatly reduces the effort required to generate high pressures. Large volume Pyrex™ reservoir can be filled during operations.