General Eastern MMY245 Portable Dewpoint Analyzer
Representative image only.
General Eastern Mfr # MMY245-RP – Item # EW-37852-16
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)-148
  • Min Temperature (° C)-100
  • Max Temperature (° F)68
  • Max Temperature (° C)20
  • Temperature Accuracy±2.0°C
  • Temperature Resolution0.1°
  • BatteryD-cell Battery (4)
  • DescriptionPortable Dewpoint Analyzer
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Convenient and portable, this moisture analyzer can quickly spot check the performance of gas dryers for compressed air, breathing air and pure gas supplies. The planar capacitive sensor combines accurate dewpoint measurement with fast response time. The sensor is stored in a very dry environment within the analyzer between measurements which speeds response time. The instrument can be configured according to the desired moisture unit of measure, temperature unit of measure, or other operating parameters.
  • Measures dewpoint within gases
  • Shoulder strap comes standard
  • Planar sensor within desiccant chamber for fastest response
  • Pressure dewpoint measurement range of -100C to +20C
  • User selectable moisture measurement units
  • Fully portable - powered by four common "D" batteries with auto shut off feature
  • Two non IS voltage outputs
  • Large 4 digit LCD display
  • Auto shut off timer (adjustable