General Resistance DA-33-3X Decade Box0.01%Acc Resistance 3 Decades
Representative image only.
General Resistance Mfr # DA-33-3X – Item # EW-20018-80
Stop guessing about optimal circuit value
  • Quickly and accurately determine the optimal value for circuit operation
  • Feel confident about measurements—accurate to ±0.01%
  • Set your applications—choose from 3 to 7 decades
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDecade Box, 0.01% Acc Resistance 3 Decades
  • Warranty1 Year
1 Year Warranty
Key Features
  • Quickly and accurately determine the optimal value for circuit operation
  • Feel confident about measurements—accurate to ±0.01%
  • Set your applications—choose from 3 to 7 decades
  • 1 year warranty
More About this Item
Choose from the Resist-O-Stat (RDS series) and Dial-An-Ohm (DA series) decade resistors. Both series are available in 3, 4, 5, 6, or 7 decades.
The RDS series resistors features Top of the line” GR performance, Rotary switches, gold-plated contacts, low contact R, and a minimum Ohms value of zero.
The DA series resistors are feature easy-to-use, economical thumbwheel switches with two mechanical configurations.
NOTE: General Resistance Decade Boxes as RTD Simulators: When the ultimate in accuracy is required, the 20019-40 or 20019-39 model should be used. These have absolute accuracy of ±0.005% and ±0.01% respectively. Due to “O” R and contact R at 100 ohms, 20019-12 can be off ±0.019%.