General Resistance RTD-100X RTD-100x, RTD Simulator, 0.005% Accuracy
Representative image only.
General Resistance Mfr # RTD-100X – Item # EW-20019-40
  • 1 year warranty
PRODUCT OPTIONS
$6,524.00USD / Each
$265.00 - $335.00USD / Each
for an additional
$335.00USD / Each
for an additional
$265.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionRTD-100x, RTD Simulator, 0.005% Accuracy
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Model RTD100
The flagship of the GR line priorto the introduction of the RTD100X, the RTD100 is identicalto the 100X except for its absolute accuracy of ± 0.01%.
Model RTD100X
The RTD100X provides an absolute accuracy of ± 0.005% over its entire resistance range. There is no need to even attempt to calculate or compensate for the error contribution due to contact resistance or "O" resistance because there is no "O" resistance. The unique shunt decadedesign reduces these errors to a level of insignificance. When the RTD100X replaces the RTD in the system, any temprature-equivalent resistance can be selected on the error-free in-line making it possible to check or calibrate all of the other elements in the system. The usable resistance range of 10 to 1111.11O is ideally suited for use with DIN PT100, PT100 ANSI, PT50 and NI120 systems.