General Tools DCS1700 Super High Performance VGA Recording Video Borescope System
Representative image only.
General Tools Mfr # DCS1700 – Item # EW-48610-75
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSuper High Performance VGA Recording Video Borescope System
  • Warranty1 Year
1 Year WarrantyConformite Europeenne
Key Features
  • 1 year warranty
More About this Item
Ideal for inspection of duct work, piping, wiring locations, and much more. Camera-tipped probes are flexible and strong at the same time. View findings instantaneously on the LCD monitor; some models will capture still and video images. Adjustable LED lighting on the probe tip provides illumination in dimly lit areas, plus allows you to lower the light intensity when viewing reflective objects to eliminate washout.
DCS1700
* Record VGA resolution (640 x 480 pixels) video clips and still images
* Probe has light boost, antireflection, and screen rotation
* Console features brightness controls, horizontal mirroring, 2X zoom, and slow-motion and fast- forward video playback
* Dual output jacks