General Tools DT80-2 Economical Type K/J Thermocouple Thermometer, Dual Input
Representative image only.
General Tools Mfr # DT80-2 – Item # EW-94461-12
Compact and lightweight design
  • Available with single or dual input
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeThermocouple
  • Number Of Probe Channels2
  • Types Of Probes AcceptedType J, Type K
  • Min Temperature (° F)-328
  • Max Temperature (° F)1922
  • Min Temperature (° C)-200
  • Max Temperature (° C)1050
  • Accuracy±0.3% of reading +2.0°F (+1.0°C); ±0.5% of reading +2.0°F (+1.0°C) above 200° and below -200°
  • Resolution0.1°
  • Height (in)5 1/4
  • No. of Probe(s) Included2
  • Battery9v Battery (1)
  • DescriptionEconomical Type K/J Thermocouple Thermometer, Dual Input
  • Warranty5 years
5 Year Warranty
Key Features
  • Available with single or dual input
  • 5 year warranty
More About this Item
These thermometers provide accurate and reliable temperature measurement in an economic and compact design. Features include min/max, Hold, °F/°C selection, relative temperature, and auto power-off after 30 minutes of inactivity to save on battery power. Accept both type K or J thermocouple probes with a miniconnector. Available in single and dual input, with results displayed on a large 1 1/4" x 1 3/4" LCD. The dual input meter (94461-12) can read and display results from two probes simultaneously and track the difference between them (T1-T2). Order optional probes and other accessories separately.
What's Included: beaded wire probe (two probes for 94461-12) and one 9 V battery.