George Ulanet 324-230 Heat-O-Matic Immersion Heater, short Rod , 5" L Heated Area, 1100 watts, 115 VAC
Representative image only.
George Ulanet Mfr # M306-02-00 – Item # EW-03046-04
Heaters with dial temperature controller
  • Plastic handle offers secure grip
  • Pilot light indicates when heater is on
  • 6 Month Product / 12 Month Parts and Labor Warranty
PRODUCT OPTIONS
SPECS & DESCRIPTION
Specifications & Description
  • Immersion Depth (in)5
  • Heated Area5 in L
  • Max Temperature (° F)212
  • Max Temperature (° C)100
  • Min TemperatureAmbient;Ambient
  • Plug TypeUS Standard
  • Power (VAC)115
  • Power (watts)1100
  • DescriptionHeat-O-Matic Immersion Heater, short Rod , 5\" L Heated Area, 1100 watts, 115 VAC
  • Warranty6 Month Product / 12 Month Parts and Labor
UL Listed to Canadian safety standards.
Key Features
  • Plastic handle offers secure grip
  • Pilot light indicates when heater is on
  • 6 Month Product / 12 Month Parts and Labor Warranty
More About this Item
Heaters with Dial Temperature Controller have a sensitivity of ±1°>F when liquid is stirred. Top-mounted pilot light indicates that heater is on. Choose from four styles "A" loop, "B" short rod, "C" flat, and "D" long rod which is the same as style "B" only longer. Heaters feature a plastic grip handle; and a 6-ft, three-wire cord, 115 VAC models have U.S. standard plug, 230 VAC models do not have a plug.