George Ulanet 492-6 Heat-O-Matic Immersion Heater with Thermostatic Controller, 1500 watts, 13" dia, 115 VAC
Representative image only.
George Ulanet Mfr # 492-6 – Item # EW-03052-50
  • 6 Month Product / 12 Month Parts and Labor Warranty
PRODUCT OPTIONS
$528.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Immersion Depth (in)5
  • Heated Area5 in L
  • Max Temperature (° F)250
  • Max Temperature (° C)121
  • Min TemperatureAmbient;Ambient
  • Plug TypeUS Standard
  • Power (VAC)115
  • Power (watts)1500
  • DescriptionHeat-O-Matic Immersion Heater with Thermostatic Controller, 1500 watts, 13\" dia, 115 VAC
  • Warranty6 Month Product / 12 Month Parts and Labor
Key Features
  • 6 Month Product / 12 Month Parts and Labor Warranty
More About this Item
Heaters with Thermostatic Temperature Controller have a sensitivity of ±2°>F(±1°>C) in an agitated bath. Heaters feature a coiled heating element to concentrate heat at bottom of vessel. A 6-ft, three-wire cord with plug is included. Voltage transformer listed below is required for 230 VAC operation.