GF Signet 161-323-551 Piping System Metering Ball Valve, PVC, 3/8" NPT(F)
Representative image only.
GF Signet Mfr # 161523521 – Item # EW-01360-40
Precise control over 180° range
  • Choose between PVC or polypropylene (PP) construction
PRODUCT OPTIONS
$126.70USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.375
  • Outlet Port3/8" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port3/8" NPT(F)
  • Seal MaterialEPDM
  • Wetted MaterialsPVC
  • DescriptionPVC Metering Ball Valve, 3/8" NPT(F)
Key Features
  • Choose between PVC or polypropylene (PP) construction
More About this Item
Ball valves give you precise control over the entire 180°> range. True union design lets you remove wetted parts. Valves have Viton® or EPDM body seals and seats. Less than 5" overall length. Calibrated in 5°> increments.