GF Signet 161.360.581 Ball Check Valve, PVC, 3/8" NPT(F); 1/Each
Representative image only.
GF Signet Mfr # 161561101 – Item # EW-01368-10
Unique square-cut elastomer seat provides seal at very low back pressures
  • Constructed of a PVC body and EPDM seal
PRODUCT OPTIONS
$57.90USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.375
  • Outlet Port3/8" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port3/8" NPT(F)
  • Cv4.8
  • Wetted MaterialsPVC
  • DescriptionPVC Ball Check Valve, 3/8" NPT(F)
Key Features
  • Constructed of a PVC body and EPDM seal
More About this Item
Quickly and easily install either vertically or horizontally. These valves are assembled with water soluble, silicone-free lubricant and constructed of a one-piece PVC body.