GF Signet 161.514.615 Type 514 PVC Manual Diaphragm Valve, 1-1/4"
Representative image only.
GF Signet Mfr # 161.514.615 – Item # EW-07390-52
Optimized valve body provides double the CV values
  • Two-color positition indicator on handwheel
  • Elimination of bolts removes corrosive weak points
  • Mount vertically or horizontally
  • PVC body and EPDM diaphragm
PRODUCT OPTIONS
$316.10USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1-1/4" true union socket
  • Connection TypeTrue union socket
  • Inlet Port1-1/4" true union socket
  • Cv51.3
  • Max Temperature (° F)150
  • Max Temperature (° C)65.5
  • Wetted MaterialsPVC, EPDM
  • Working Pressure (PSI)150
  • DescriptionType 514 PVC Manual Diaphragm Valve, 1-1/4"
Key Features
  • Two-color positition indicator on handwheel
  • Elimination of bolts removes corrosive weak points
  • Mount vertically or horizontally
  • PVC body and EPDM diaphragm
More About this Item
These GF Signet® manual diaphragm valves are constructed with a PVC body and EPDM diaphragm. Valves offer a maximum temperature of 150°F (65°C) and feature true union socket connections.