GF Signet 161-523-522 Piping System Metering Ball Valve, PVC, 1/2" NPT(F)
Representative image only.
GF Signet Mfr # 161523522 – Item # EW-01360-41
Precise control over 180° range
  • Choose between PVC or polypropylene (PP) construction
PRODUCT OPTIONS
$130.30USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.5
  • Outlet Port1/2" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/2" NPT(F)
  • Seal MaterialEPDM
  • Wetted MaterialsPVC
  • DescriptionPVC Metering Ball Valve, 1/2" NPT(F)
Key Features
  • Choose between PVC or polypropylene (PP) construction
More About this Item
Ball valves give you precise control over the entire 180°> range. True union design lets you remove wetted parts. Valves have Viton® or EPDM body seals and seats. Less than 5" overall length. Calibrated in 5°> increments.