GF Signet 161.546.344 Type 546 Ball Valve, PVC with EPDM seals, 1"
Representative image only.
GF Signet Mfr # 161.546.344 – Item # EW-01357-07
Optimize safety with this true union ball valve
  • True union ball valve with added safety and functionality for a diverse range of applications
  • PTFE ball seat with high abrasion resistance
  • Built-in tool for easy adjustment of ball torque
  • Threaded mounting inserts for fixed-point attachment
PRODUCT OPTIONS
$120.30USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1" NPT(F) and socket
  • Connection TypeNPT(F) and Socket
  • Inlet Port1" NPT(F) and socket
  • Cv49
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • Wetted MaterialsPVC, EPDM
  • Working Pressure (PSI)Socket: 232 / Thread: 145
  • DescriptionType 546 Ball Valve, PVC with EPDM seals, 1"
Key Features
  • True union ball valve with added safety and functionality for a diverse range of applications
  • PTFE ball seat with high abrasion resistance
  • Built-in tool for easy adjustment of ball torque
  • Threaded mounting inserts for fixed-point attachment
More About this Item
Ball valves are designed primarily for on/off service. These valves contain a ball with a hole through it. A handle or electronic actuator rotates the ball 90 degrees, turning the flow on or off.