GF Signet 163.546.353 Type 546 Ball Valve, CPVC with seals, 3/4"
Representative image only.
GF Signet Mfr # 163.546.353 – Item # EW-01371-10
Optimize safety with this true union ball valve
  • True union ball valve with added safety and functionality for a diverse range of applications
  • PTFE ball seat with high abrasion resistance
  • Built-in tool for easy adjustment of ball torque
  • Threaded mounting inserts for fixed-point attachment
PRODUCT OPTIONS
$162.90USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port3/4" NPT(F) and socket
  • Connection TypeNPT(F) and Socket
  • Inlet Port3/4" NPT(F) and socket
  • Cv24.5
  • Max Temperature (° F)176
  • Max Temperature (° C)80
  • Wetted MaterialsCPVC, Viton®
  • Working Pressure (PSI)Socket: 232 / Thread: 145
  • DescriptionType 546 Ball Valve, CPVC with seals, 3/4"
Key Features
  • True union ball valve with added safety and functionality for a diverse range of applications
  • PTFE ball seat with high abrasion resistance
  • Built-in tool for easy adjustment of ball torque
  • Threaded mounting inserts for fixed-point attachment
More About this Item
Ball valves are designed primarily for on/off service. These valves contain a ball with a hole through it. A handle or electronic actuator rotates the ball 90 degrees, turning the flow on or off.