GF Signet 3-2350-1 Remote Temperature Sensor, 15 ft Cable
Representative image only.
GF Signet Mfr # 3-2350-1 – Item # EW-93310-00
  • RTD sensors are threaded on both ends for submersion in process vessels or in-line applications
  • Use the remote sensor with the field or panel mount transmitter to display temperatures anywhere.
  • 2 year warranty
$607.00USD / Each
$209.00 - $279.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Process Connection3/4"
  • Temperature Accuracy±0.9°F (0.5°C)
  • DescriptionRemote Temperature Sensor, 15 ft Cable
  • Warranty2 year
Two year warranty
Key Features
  • RTD sensors are threaded on both ends for submersion in process vessels or in-line applications
  • Use the remote sensor with the field or panel mount transmitter to display temperatures anywhere.
  • 2 year warranty
More About this Item
Sensors feature a one-piece body made from PVDF for purity and durability. Available with either a 15-ft or 6'' cable; extend cable up to 200-ft by using two-conductor shielded cable.