GF Signet 3-2536-P1 Rotor-X Flow Sensor, PP/Ti/PVDF for 5-8" Pipe
Representative image only.
GF Signet Mfr # 3-2536-P1 – Item # EW-32500-02
Measure low flow rates down to 0.3 ft/second
  • Eliminate cavitation and minimize pressure drop
PRODUCT OPTIONS
$543.50USD / Each
$310.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Repeatability±0.5% full-scale
  • Wetted MaterialsPolypropylene, Titanium, PVDF
  • Max Operating Temperature (° F)185
  • Max Operating Temperature (° C)85
  • Pipe Size5 to 8'
  • Power5 to 24 VDC
  • DescriptionRotor-X Flow Sensor, PP/Ti/PVDF for 5-8" Pipe
Key Features
  • Eliminate cavitation and minimize pressure drop
More About this Item
Measure low flow rates in almost any size pipe. These sensors contain an open-core rotor with a small magnet in each of the four blades. As they rotate, the magnets pass a Hall effect sensor, which generates a linear frequency output that is proportional to the flow velocity. This signal travels up to 1000 feet without amplification.
The open-core design eliminates cavitation at flow velocities from 0.3 to 20 ft/second. Closed or solid paddle-wheel designs create air bubbles at these velocities, causing a nonlinear and nonrepeatable signal.
Sensor materials resist most chemicals. All models have PVDF rotor and Viton® O-ring; and include a 25-ft connecting cable.
ROTOR-X low-flow sensors are compatible with all of our line-powered flow indicators, accumulators, and controllers. Mount in any type of pipe. Order installation fittings separately.