GF Signet 3-2760-2 DryLoc Preamp with ISO threads, 15-ft cable
Representative image only.
GF Signet Mfr # 3-2760-2 – Item # EW-56560-04
$438.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ConnectorDryLoc (ISO 7/1 R3/4)
  • Cable Length (m)4.5
  • DescriptionDryLoc Preamp with ISO threads, 15-ft cable
More About this Item
These preamplifiers will allow any DryLoc pH/ORP electrodes to work with most +GF+ SIGNET instruments. These devices will self-configure for pH or ORP operation via automatic recognition of the electrode type. Electrodes with EasyCal allow simple push-button calibration and include an LED indicator for visual feedback.
The DryLoc electrode connector forms a robust assembly for submersible and in-line installations. NEMA 4X junction boxes are integral parts of in-line versions and are available as accessories for submersible versions.
Call Us At1-847-549-7600
Live chat