GF Signet Installation Fitting, Tee, Carbon Steel, 1" Pipe ID
Representative image only.
GF Signet Mfr # CS4T010 – Item # EW-32511-74
Build a complete paddle-wheel flow sensor system customizable to your application
  • Modular system for a customizable approach to flow measurement
  • Ideal for easy installation
PRODUCT OPTIONS
$421.20USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Material FittingCarbon Steel
  • Process ConnectionNPT Thread
  • Pipe Size1"
  • DescriptionInstallation Fitting, Tee, Carbon Steel, 1" Pipe ID
Key Features
  • Modular system for a customizable approach to flow measurement
  • Ideal for easy installation
More About this Item
The GF Signet paddle-wheel flow system is comprised of three components. Select the flow sensor, installation fitting, and flow meter that suit your application needs for a customizable approach to flow measurement.
Step One: Choose a Flow Sensor
Step Two: Choose an Installation Fitting
Step Three: Choose a Flow Meter
Flow sensor, installation fitting, and flow meter sold separately.