GF Signet Installation Fitting, Weldolet Tee, Carbon Steel, 8" Pipe ID
Representative image only.
GF Signet Mfr # CS4W080 – Item # EW-32511-83
Build a complete paddle-wheel flow sensor system customizable to your application
  • Modular system for a customizable approach to flow measurement
  • Ideal for easy installation
PRODUCT OPTIONS
$276.20USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Material FittingCarbon Steel
  • Process ConnectionWeldolet
  • Pipe Size8"
  • DescriptionInstallation Fitting, Weldolet Tee, Carbon Steel, 8" Pipe ID
Key Features
  • Modular system for a customizable approach to flow measurement
  • Ideal for easy installation
More About this Item
The GF Signet paddle-wheel flow system is comprised of three components. Select the flow sensor, installation fitting, and flow meter that suit your application needs for a customizable approach to flow measurement.
Step One: Choose a Flow Sensor
Step Two: Choose an Installation Fitting
Step Three: Choose a Flow Meter
Flow sensor, installation fitting, and flow meter sold separately.