GF Signet Metal Installation Tee Fitting, 316 Stainless Steel, 3/4" Pipe ID
Representative image only.
GF Signet Mfr # CR4T007 – Item # EW-05620-42
Ensure proper insertion depth and measurement
  • Choose from 316 Stainless Steel (SS), carbon steel (CS), galvanized iron (schedule 40), and copper tee fittings
PRODUCT OPTIONS
$588.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting Material316 Stainless Steel
  • Connection TypeNPT(F)
  • Thread size NPT(F) (port 1) (in)3/4
  • Thread size NPT(F) (port 2) (in)3/4
  • DescriptionMetal Installation Tee Fitting, 316 Stainless Steel, 3/4" Pipe ID
Key Features
  • Choose from 316 Stainless Steel (SS), carbon steel (CS), galvanized iron (schedule 40), and copper tee fittings
More About this Item
These metal installation tee fittings are available in 316 Stainless Steel (SS), carbon steel (CS), galvanized iron (schedule 40), and copper materials in a wide range of sizes from 3/4” to 8” pipe inner diameters.
Standard metal tees constructed of SS, CS, and galvanized iron fittings include PVDF insert for all sizes and NPT(F) threads at each end. Copper tee fittings include PVDF insert for pipe ID over 1" and feature sweat-on ends.
Weldolet CS fittings weld directly onto hole cut in pipe.