GF Signet Metal Installation Tee Fitting, Strap-On Saddle , PVC 80, 6" Pipe ID
Representative image only.
GF Signet Mfr # PVUS060 – Item # EW-05620-83
Ensure proper insertion depth and measurement
  • Constructed of durable PVC 40 or PVC 80
PRODUCT OPTIONS
$358.60USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPVC 80
  • Connection TypeSaddle
  • Orifice Dimensions (in)6
  • DescriptionMetal Installation Tee Fitting, Strap-On Saddle , PVC 80, 6" Pipe ID
Key Features
  • Constructed of durable PVC 40 or PVC 80
More About this Item
These PVC saddle installation fittings are available for PVC 40 or PVC 80 pipes from 2” to 8” diameters.