GF Signet P526-2007 316 SS Socket-Weld Mini-Tap Fittings; 3/4" Pipe ID
Representative image only.
GF Signet Mfr # P526-2007 – Item # EW-05618-62
Installation fittings for your GF® Signet Paddle Wheel Flowmeters
  • 316 SS Socket-Weld Mini-Tap Fittings for Metalex™ sensors only
  • Includes cap kit
PRODUCT OPTIONS
$621.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Wetted Materials316 Stainless Steel
  • Pipe Size0.75"
  • Description316 SS Socket-Weld Mini-Tap Fittings; 3/4" Pipe ID
Key Features
  • 316 SS Socket-Weld Mini-Tap Fittings for Metalex™ sensors only
  • Includes cap kit
More About this Item
Fittings are precision crafted to ensure proper sensor insertion depth and accurate flow measurement. Pipe fitting sizes range from 1/2" to 1" in diameter.
Required system components are the flow sensor, installation fitting, and monitor or controller, all sold separately.