GF Signet P526-2080 Metalex Mini-Tap, weld-on, 8" pipe
Representative image only.
GF Signet Mfr # P526-2080 – Item # EW-32511-95
$427.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Process ConnectionWeld-on
  • Pipe Size8"
  • DescriptionMetalex Mini-Tap, weld-on, 8" pipe
More About this Item
Stainless steel body withstands high pressures and temperatures Use these rugged sensors to monitor the flow of liquid ammonia, turbine steam condensate, and more - wherever strength and chemical resistance are required.
Step One: Choose a Flow Sensor
Step Two: Choose an Installation Fitting
Step Three: Choose a Flow Meter