GF Signet PV8S030 PVC Strap-On Saddle for 3" PVC 40/80 Pipe
Representative image only.
GF Signet Mfr # PV8S030 – Item # EW-05620-18
$224.70USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPVC 40
  • Connection TypeSaddle
  • DescriptionPVC Strap-On Saddle for 3" PVC 40/80 Pipe
More About this Item
PVC Saddle Fittings for PVC 40 pipes from 2" to 4" in diameter; PVC 80 pipes from 6" to 8" in diameter. Specify schedule of pipe when ordering.