GF Signet PV8T020 Installation Fittings, tee pvc80 for 2" ID pipe
Representative image only.
GF Signet Mfr # MPV8T020 – Item # EW-05620-26
$288.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPVC 80
  • Connection TypeSlip fit
  • DescriptionInstallation Fittings, tee pvc80 for 2" ID pipe
More About this Item
PVC Tee Fittings for PVC 80 pipes up to 4" in diameter and CPVC 80 pipes up to 1 1/2" in diameter. Slip on ends (no threads).sensors.