GF Signet TYPE-546 Type 546 Ball Valve, Manual, PP, 1/2"NPT(F), FPM Seals
Representative image only.
GF Signet Mfr # 175.546.332(TYPE-546) – Item # EW-01374-92
Optimize safety with this true union ball valve
  • True union ball valve with added safety and functionality for a diverse range of applications
  • PTFE ball seat with high abrasion resistance
  • Built-in tool for easy adjustment of ball torque
  • Threaded mounting inserts for fixed-point attachment
PRODUCT OPTIONS
$635.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1/2" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/2" NPT(F)
  • Cv13
  • Max Pressure (PSI)145
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • Wetted MaterialsPVDF, PTFE, FPM
  • DescriptionType 546 Ball Valve, Manual, PP, 1/2"NPT(F), FPM Seals
Key Features
  • True union ball valve with added safety and functionality for a diverse range of applications
  • PTFE ball seat with high abrasion resistance
  • Built-in tool for easy adjustment of ball torque
  • Threaded mounting inserts for fixed-point attachment
More About this Item
Ball valves are designed primarily for on/off service. These valves contain a ball with a hole through it. A handle or electronic actuator rotates the ball 90 degrees, turning the flow on or off.