GF Signet Weld-On Installation Fitting, 316 Stainless Steel, 1-1/4" Mini-Tap Pipe ID
Representative image only.
GF Signet Mfr # P526-2012 – Item # EW-05618-65
Ensure proper insertion depth and measurement
  • Choose from a wide range of pipe diameters to suit your application
PRODUCT OPTIONS
$427.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting Material316 Stainless Steel
  • Connection TypeWeld-On Mini-Tap
  • Orifice Dimensions (in)1 1/4
  • DescriptionWeld-On Installation Fitting, 316 Stainless Steel, 1-1/4" Mini-Tap Pipe ID
Includes
cap kit.
Key Features
  • Choose from a wide range of pipe diameters to suit your application
More About this Item
These GF Signet 316 Stainless Steel weld-on mini-tap installation fittings are suitable for use with Metalex sensors only. All fittings include a cap kit.