Gilmont Falling Ball Viscometer, 0.2 to 10 cP

Representative image only.
Gilmont Item # EW-08701-00
Gilmont viscometers are ideal for routine plant quality control checks and research laboratory applications

  • Viscometer features
  • Easy use design
  • Viscosity range (cP): 0.2 to 10
  • High precision falling ball

PRODUCT OPTIONS

$326.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Min Viscosity Range (cP)0.2
  • Max Viscosity Range (cP)10
  • Sample Volume Needed (mL)7
  • DescriptionFalling Ball Viscometer, 0.2 to 2 / 1 to 10 centipoise

Includes

One glass and one 316 stainless steel (SS) high-precision ball. For higher viscosity ranges, use the optional tantalum ball (sold separately).

Key Features


  • Viscometer features
  • Easy use design
  • Viscosity range (cP): 0.2 to 10
  • High precision falling ball

More About this Item

This viscometer is extremely easy to use; first fill the tube with sample, then invert tube. Next, restore tube to normal position, turn the knob to release the ball, and then simply measure the time of descent. The ball moves against a white background with red reference lines making it easy to read. Repeatability with good technique varies from ±0.2 to ±1.0%, depending upon time of descent. Falling ball viscometers require a 7-mL sample volume.
Gilmont viscometers have wetted materials of glass, Viton®, and Delrin® acetal. PTFE O-rings are available for use with lquids that adhere to Viton®; call our Applications Specialists for more information.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat