This item is now discontinued. Please browse similar items
Gilmont Falling Ball Viscometer, 20 to 200 / 100 to 1000 centipoise
Representative image only.
Gilmont Item # EW-08702-10
Gilmont viscometers are ideal for routine plant quality control checks and research laboratory applications
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Min Viscosity Range (cP)20
  • Max Viscosity Range (cP)1000
  • Sample Volume Needed (mL)7
  • DescriptionFalling Ball Viscometer, 20 to 200 / 100 to 1000 centipoise
Includes
One glass and one 316 stainless steel (SS) high-precision ball. For higher viscosity ranges, use the optional tantalum ball (sold separately).
More About this Item
This viscometer is extremely easy to use; first fill the tube with sample, then invert tube. Next, restore tube to normal position, turn the knob to release the ball, and then simply measure the time of descent. The ball moves against a white background with red reference lines making it easy to read. Repeatability with good technique varies from ±0.2 to ±1.0%, depending upon time of descent. Falling ball viscometers require a 7-mL sample volume.
Gilmont viscometers have wetted materials of glass, Viton®, and Delrin® acetal. PTFE O-rings are available for use with lquids that adhere to Viton®; call our Applications Specialists for more information.