Gilmont GF-1360 Shielded Correlated Flowmeter, Tube Size 3
Representative image only.
Gilmont Item # EW-03203-20
  • Glass float also included: when glass float is used flow ranges are approximately half the values listed (for 316SS)
PRODUCT OPTIONS
$473.00USD / Each
$262.00USD / Each
for an additional
$262.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±2% of reading or ±1 scale division; whichever is greater
  • Max Pressure (PSI)75
  • Max Air Flow Rate (mL/min)25000
  • Max Water Flow Rate (mL/min)640
  • Min Flow Rate (mL/min)200
  • Max Flow Rate (mL/min)640
  • Material FloatGlass or 316 SS
  • Max Operating Temperature (° F)150
  • Max Operating Temperature (° C)65
  • Pipe Size3
  • Fluid TypeWater
  • Media TypeCarbon Dioxide;Carbon Monoxide;Oxygen;Acetylene;Air;Argon;Hydrogen;Water;Nitrogen;Helium
  • Gas TypeAir
  • DescriptionShielded Correlated Flowmeter, Tube Size 3
Key Features
  • Glass float also included: when glass float is used flow ranges are approximately half the values listed (for 316SS)
More About this Item
Correlated Flowmeters
Extremely accurate—±2% of reading or ±1 scale division, whichever is greater. Flowmeters with micro-sized tubes (models 03210-00, 03210-20, and 03234-50) are accurate to ±5% of reading or ±2% of scale whichever is greater. Computer calibrated flow charts are included for floats used with both air and water at STP. An “R factor” chart and formulas are included to convert scale readings for other gases or liquids or for floats other than glass. Order our flow analysis software below to generate flow charts specific to your individual applications.
Shielded models accept 3/8" ID tubing (sizes 4 and 5 accept 5/8" ID tubing). PTFE coupling adapters for unshielded models are not included—order separately.
ACCESSORIES
REVIEWS
Q&A
Do you have questions about this product?
Get answers from real customers and in-house experts with AnswerBox.
1 Question | 1 Answer
Displaying question 1
  • Q:
    Good Day,
    I have a Gilmont Shielded Flowmeter Size 3 model number GF-1360. I need polypropylene barb fittings for this flowmeter that can be used with 3/8" ID tubing are they available? If so what is the price?
    Thanks,
    Asked on 4/13/2011 by Heidi Gaebler from Fort Collins, CO
    1 answer
    • STAFF EXPERT
      A:
      This flowmeter has 1/4" NPT male threads. The fitting that will fit your needs best is our 31702-22, except this is Nylon, not polypropylene.
      Answered on 4/15/2011 by Josh from Technical Support
Displaying question 1
Do you have a question about this product? 