This item is now discontinued. Please browse similar items
Gilmont GS-1101-A micrometer burette tip, 0.2 mL, for use with 07842-00
Representative image only.
Gilmont Item # EW-07843-00
Dispense small volumes at constant velocity—ideal for titrations and fluid injections
  • Autoclavable
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)0.2
  • Graduation Interval (mL)0.0002
  • Descriptionmicrometer burette tip, 0.2 mL, for use with 07842-00
Key Features
  • Autoclavable
More About this Item
Turn the handle of this dispenser for precise delivery of small volumes. Dispense fluids with an accuracy of ±0.5% of reading or ±1 scale division, whichever is greater. Fluids only come in contact with the borosilicate glass tip, PTFE syringe barrel, and Viton® O-rings. Optional PTFE O-rings are available separately for applications working with harsh chemicals.