Gilmont GS-1200 Micrometer Syringe, 2.0 mL; 1/Ea
Representative image only.
Gilmont Item # EW-07844-00
Dispense small volumes at constant velocity—ideal for titrations and fluid injections
PRODUCT OPTIONS
$288.20USD / Each
$76.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)2
  • Graduation Interval (mL)0.002
  • DescriptionMicrometer Syringe, 2.0 mL; 1/Ea
More About this Item
Turn the handle of this dispenser for precise delivery of small volumes. Dispense fluids with an accuracy of ±0.5% of reading or ±1 scale division, whichever is greater. Fluids only come in contact with the borosilicate glass tip, PTFE syringe barrel, and Viton® O-rings.
Autoclavable
Syringe-style pipettes terminate in a standard luer-lock fitting for easy replacement and interchangeability. Dispensers include one tip.