Gilmont Unshielded Flowmeter Size 11 Tube
Representative image only.
Gilmont Item # EW-03230-11
Design makes each meter suitable for a broad range of laboratory applications
  • Modular components allow for meter flexibility across a wide range of flow rates
  • Multiple end connections to suit process requirements
  • Accepts 5⁄16" ID tubing
  • Glass taper joints are an alternate connection option which slip on the PTFE stops that contain the float (contact our Applications Specialists to order the glass taper joints).
PRODUCT OPTIONS
$214.00USD / Each
$262.00USD / Each
for an additional
$262.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy0.05% of reading or 2 mm of the scale length, whichever is greater
  • Max Air Flow Rate (mL/min)500
  • Max Water Flow Rate (mL/min)8.6
  • Max Flow Rate (mL/min)500
  • Material Flow TubeBorosilicate glass
  • Material FittingPTFE
  • Material ValveGlass chamber with PCTFE plug
  • Material O-RingViton®
  • Material FloatGlass or 316 SS
  • Material HousingPC
  • Max Operating Temperature (° F)150
  • Max Operating Temperature (° C)65
  • Fluid Typeair and water
  • Media TypeHelium;Hydrogen;Nitrogen;Argon;Carbon Dioxide;Carbon Monoxide;Oxygen;Acetylene;Air;Water
  • Gas TypeAir
  • Display TypeSize 11 Tube
  • DescriptionUnshielded Flowmeter Size 11 Tube
Key Features
  • Modular components allow for meter flexibility across a wide range of flow rates
  • Multiple end connections to suit process requirements
  • Accepts 5⁄16" ID tubing
  • Glass taper joints are an alternate connection option which slip on the PTFE stops that contain the float (contact our Applications Specialists to order the glass taper joints).
More About this Item
Choose from unshielded, shielded, or shielded with valve flow meters suitable for a wide range of applications.
Unshielded Flowmeters work well in low-pressure applications. Connect the flowtube end directly to tubing having the proper inner diameter. Maximum pressure rating for unshielded flowmeters is 15 psi.
Shielded Flowmeters are better suited to higher-pressure applications or for installations requiring a panel-mounted flowmeter. End bushings are molded with dual connection capability—tubing or threaded.
Shielded Flowmeters with Valves offer flow monitoring along with precise control through an integrated 20-turn micrometer valve. The valve can be adjusted from 0.1 to 100% of maximum flow (semilogarithmic) and provides recise regulation from 0.3 to 60% of maximum flow.
The tube size 11 flowmeter has a flow range for air of 1 to 280 mL/min and 0.01 to 4.0 mL/min for water with glass float.
Glass taper joints are available upon request—call our Application Specialists to order.