Glas-Col 100AO812 Buchner Funnel Mantle for Coors Funnel No. 60243, 70 Watts, 115 V

Representative image only.
Glas-Col Mfr # 100AO812 – Item # EW-36227-68
Get better heating with conforming flexible fabric
  • 1 year warranty

PRODUCT OPTIONS

$723.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Coors Funnel Number60243
  • MaterialFabric
  • Max temperature (° F)842
  • Max temperature (° C)450
  • Power (VAC)115
  • Power (watts)70
  • Diameter (in)3.25
  • Outside Diameter (OD) (in)5.75
  • DescriptionBuchner Funnel Mantle for Coors Funnel No. 60243, 70 Watts, 115 V
  • Warranty1 year
1 Year Warranty

Key Features

  • 1 year warranty

More About this Item

Heating Mantles feature insulated glass fabric that conforms to unusual vessel shapes. Büchner Funnel Mantles provide gentle heat for solutions that must be filtered while warm. Mantles require a controlled electrical power source to prevent overheating.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat