Hach 3726E2T 1000 Ohm electrodeless conductivity sensor, PVDF, 3/4
Representative image only.
Hach Mfr # 3726E2T – Item # EW-19901-00
Innovative technology eliminates polarization and electrode coating problems in harsh environments
  • Rugged, Low maintenance, non-fouling design
  • Available in polpropylene, PVDF, PEEK, or PFA wetted materials
  • Standard 20-ft integral cable
  • 1 year warranty
$1,498.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Cell TypeToroidal
  • Body MaterialPVDF
  • Fitting¾\" NPT(M)
  • Mounting TypeThreaded
  • Min temperature (° C)-5
  • Max temperature (° C)200
  • Min temperature (° F)23
  • Max temperature (° F)392
  • Use With19065-24, -34, -44
  • Cable Length (m)6
  • Cable Length (ft)20
  • Description1000 Ohm electrodeless conductivity sensor, PVDF, 3/4
  • Warranty1 year
1 Year Warranty
Key Features
  • Rugged, Low maintenance, non-fouling design
  • Available in polpropylene, PVDF, PEEK, or PFA wetted materials
  • Standard 20-ft integral cable
  • 1 year warranty
More About this Item
Sensors induce a low current in a closed loop of solution, then measures the magnitude of this current to determine a solutions conductivity. Problems such as polarization, oily fouling, process coating, and contamination will not affect the performance of the electrodeless sensor.
Sensors are constructed of only one wetted material—choose from polypropylene (PP), PVDF, PEEK, or PFA. These nonconductive materials ensure the cell is electrically isolated from the process fluid. This eliminates ground loops which can affect accuracy.
Sensors measure from 0 to 200 µS up to 0 to 2,000,000 µS at a maximum flow rate of 10 ft per second. Cells include automatic internal temperature compensation (32 to 257°F, 0 to 125°C), and a 20 ft, PTFE jacketed, five conductor integral cable rated up to 200°C.