Global Cooling ULT25-120 Shuttle Portable Ultra-Low Temperature Freezer, 120/220V
Representative image only.
Global Cooling Mfr # ULT25N-000000 – Item # EW-44199-05
Energy-efficient ultra-low temperature storage anywhere you need it!
  • Weighs less than 42 lb (19 kg), making it ideal for benchtop or field applications
  • Easy-grip molded-in handles offer comfortable transport
  • A change from AC to DC operation is as easy as changing the power cord (power cord sold separately)
  • Connects easily to a mobile power supply
$7,044.00USD / Each
$205.00USD / Each
for an additional
$205.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Freezer Capacity (cu ft)3.1
  • Freezer Capacity (Liters)88
  • Max Temperature - Freezer (° C)-50
  • Power (VAC)220
  • Power (Hz)50
  • Power (Breaker)15 A
  • DescriptionShuttle Portable Ultra-Low Temperature Freezer, 120/220V
Includes
one plastic utility bin.
Key Features
  • Weighs less than 42 lb (19 kg), making it ideal for benchtop or field applications
  • Easy-grip molded-in handles offer comfortable transport
  • A change from AC to DC operation is as easy as changing the power cord (power cord sold separately)
  • Connects easily to a mobile power supply
More About this Item
This lighweight portable freezer was developed to bring ultra-low -86°C temperature storage to places conventional compressor freezers cannot go or won’t work. It offers secure mobile transport of biological specimens. Ideal for use within biorepositories, and other applications associated with pharmaceutical distribution.
Operating on a stirling engine, the freezer requires no compressors, dry ice, or LN2, and draws the same amount of energy as a conventional light bulb, making it environmentally friendly and cost effective. The Humm™ free-piston stirling engine contains two moving parts that float on helium gas bearings to eliminate contact wear. The linear driver modulates engine power to deliver cooling on demand for straight line control without on/off cycling. Freezing is performed by the thermosiphon evaporator. A microcharge of environmentally friendly cooling medium creates a uniform ultra-low temperature by a continuous process of evaporation and condensation within the sealed tube.
Weather-resistant touchpad controller features a digital temperature display indicating interior temperature and error codes if detected. Temperature preset buttons allow you to quickly choose -20, -40, or -86°C (default at startup); easily raise and lower temperature at 1°C intervals. A system lock indicator confirms system set points are locked and prevents errors or unauthorized adjustment. The over/under-temperature alarm activates visual and audible beep warnings if temperature exceeds set point and silences automatically when set point is restored.
Rugged cabinet design withstands travel inside and outside your facility. Composite vacuum-insulation panels with CFC-free foam strengthen and protect from high ambient temperatures. A double gasket seals the interior chamber enabling the lid to latch and provides dual protection against moisture and heat gain. Inner and outer gaskets are easily replaced in the field without tools. A sub-lid constructed of high-density, closed-cell rigid foam enhances temperature uniformity and provides additional protection when lid is opened. Molded-in handles make the freezer easy to grip and move.
Other features include a slide-out air filter that is easy to remove and clean, and a 1/4" (0.64 cm) access port with stopper.