Global Water PL200-G garden hose pressure logger

Representative image only.
Global Water Mfr # PL200-G – Item # EW-32922-70
Standard ¾” garden hose connection and battery operation let you record water pressure just about anywhere
  • Fast datalogging capabilities easily catch pressure dips and spikes
  • USB data port and bundled software allow for easy setup, calibration, and data transfer
  • Water-resistant enclosure and rugged housing provide for long service life

$653.25USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Number of Channels1
  • Input TypePressure
  • Max Pressure200 psi
  • Min Pressure0 psi
  • Pressure Accuracy±1%
  • Memory Size81,759 readings
  • Battery Type2 x 9V batteries
  • Length (in)4
  • Height (in)3
  • Width (in)3
  • Length (cm)10
  • Height (cm)8
  • Width (cm)8
  • Descriptiongarden hose pressure logger

Includes

software, USB cable, and two 9 V batteries. Full specifications can be found in the manual linked below

Key Features

  • Fast datalogging capabilities easily catch pressure dips and spikes
  • USB data port and bundled software allow for easy setup, calibration, and data transfer
  • Water-resistant enclosure and rugged housing provide for long service life

More About this Item

Garden hose pressure logger lets you easily identify and record water pressure dips and spikes for advanced water distribution system modeling. Compact, water-resistant NEMA 4X housing handles harsh environments and offers long service life.

Use the flexible datalogging capabilities to sample pressure at defined intervals, high speeds, logarithmically, or after programmed deviations from earlier readings. Massive, nonvolatile flash memory buffer stores over 81,000 readings. Programmable stop and start alarm times let you synchronize multiple units, delay starting until a preset time, or limit the number of recordings in a day. Bundled software exports readings via a USB data port to your computer. Software additionally provides for easy setup, calibration, and data management.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat