Global Water WL16U-060-075 Water-Level Logger, 60 foot range with 75 foot cable
Representative image only.
Global Water Mfr # WL16U-060-075 – Item # EW-32922-62
Datalogging capabilities let you remotely monitor and record water levels
  • Ideal for use with ground water, surface water, storm water, and sewers
  • Temperature- and barometric-pressure compensated readings with vented cable
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWater-Level Logger, 60 foot range with 75 foot cable
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Ideal for use with ground water, surface water, storm water, and sewers
  • Temperature- and barometric-pressure compensated readings with vented cable
  • 1 year warranty
More About this Item
These highly accurate waterlevel loggers can record over 81,000 readings. Program the logger to store data with one of four programs: fast (10 measurements per second), programmable interval (1 second to multiple years), logarithmic, and exception. Cable lengths up to 500 feet and serial communications models available; contact our Application Specialists for details.
Units include Windows®-based software for easy option setting, stored data access, and data download into a standard spreadsheet. The software also provides additional features such as real-time monitoring, measurement interval with start and stops, engineering unit selections, station ID, and sensor calibration. USB connections provide convenient connection for downloads without having to remove the sensor from the well.
Weatherproof loggers easily mount to a 2'' wellhead with standard hardware. The internal 9 V batteries power the logger and sensor for up to a year, depending on the recording interval. A third battery retains data even if the other two batteries fail.