Global Water WSSWK Stormwater kit for samplers. With rain sensor, flow sensor, and autodrain rain gauge
Representative image only.
Global Water Mfr # CTU000 – Item # EW-99243-12
Optimize water sampling
  • A variety of water sampler accessories to choose from
  • 1 year warranty
PRODUCT OPTIONS
$465.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionStormwater kit for samplers. With rain sensor, flow sensor, and autodrain rain gauge
  • Warranty1 year
1 Year Warranty
Key Features
  • A variety of water sampler accessories to choose from
  • 1 year warranty
More About this Item
Update, upgrade, or replace sampling system parts and accessories.
Stormwater kit includes a rain sensor, flow sensor, and rain guage in a single package to perform stormwater sampling when connected to a Global Water peristalic sampler.