GoatThroat SO12 12in Threaded Standoff Kit For 55 Gallon Drums
Representative image only.
GoatThroat Mfr # SO12 – Item # EW-75650-68
Enhance versitility of your GoatThroat™ Spillproof Barrel Pump
PRODUCT OPTIONS
$62.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Description12in Threaded Standoff Kit For 55 Gallon Drums
More About this Item
Accessories for GoatThroat Spillproof Barrel Pump optimize performance or suitable for maintenance.
The pneumatic air adapter, model 75650-40, allows GoatThroat™ pumps to be used with shop air, air compressors, or bottle compressed air. Uses up to 4 psi to deliver fluids up to 4.5 GPM. External pressure valve keeps containers from being over pressurized.