Graphic Controls 00013151 Circular Chart, 7 day, 12" dia, 0-100 & 0-250 scales
Representative image only.
Graphic Controls Mfr # 00013151 – Item # EW-49000-46
Get exacting performance that meets American Gas Association (AGA) standards
  • Calibrate pen arrays to a 15-minute time lag between static and differential on the Zero Line of a 24 hour chart
  • Optimal chart paper thickness prevents curling
  • Welker Slot designed for use with automatic chart changers
  • Scales read 0 to 100 and 0 to 250
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Chart TypeCircular
  • Chart Speed7 day
  • Chart Diameter (in)12
  • DescriptionCircular Chart, 7 day, 12\" dia, 0-100 & 0-250 scales
  • Warranty1 year
1 Year Warranty
Key Features
  • Calibrate pen arrays to a 15-minute time lag between static and differential on the Zero Line of a 24 hour chart
  • Optimal chart paper thickness prevents curling
  • Welker Slot designed for use with automatic chart changers
  • Scales read 0 to 100 and 0 to 250
  • 1 year warranty
More About this Item
Use with American and Barton chart recorders.