Graphic Controls 00379990 Circular Chart, Mercury Instrument, 10 hour
Representative image only.
Graphic Controls Mfr # 00379990 – Item # EW-49000-63
Get exacting performance that meets American Gas Association (AGA) standards
  • Calibrate pen arrays to a 15-minute time lag between static and differential on the Zero Line of a 24 hour chart
  • Optimal chart paper thickness prevents curling
  • Welker Slot designed for use with automatic chart changers
  • Scale reads 0 to 100
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Chart TypeCircular
  • Chart Speed10 hour
  • Chart Diameter (in)8
  • DescriptionCircular Chart, Mercury Instrument, 10 hour
  • Warranty1 year
1 Year Warranty
Key Features
  • Calibrate pen arrays to a 15-minute time lag between static and differential on the Zero Line of a 24 hour chart
  • Optimal chart paper thickness prevents curling
  • Welker Slot designed for use with automatic chart changers
  • Scale reads 0 to 100
  • 1 year warranty
More About this Item
Use with Mercury chart recorders.